Assay Details
| Gene Symbol: Cngb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271238.1 | NP_001258167.1 | 3 - 4 | 315 | 102 | |
| XM_008763596.1 | XP_008761818.1 | 1 - 2 | 559 | 102 | ||
| GenBank mRNA | AY564232.1 | - | 3 - 4 | 252 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cngb3
Genomic Map