Assay Details
| Gene Symbol: Car9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107956.1 | NP_001101426.1 | 1 - 2 | 390 | 77 | |
| XM_008763676.1 | XP_008761898.1 | 1 - 2 | 1046 | 77 | ||
| GenBank mRNA | EF187254.1 | - | 1 - 2 | 392 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car9
Genomic Map