Assay Details
| Gene Symbol: Car9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107956.1 | NP_001101426.1 | 3 - 4 | 583 | 54 | |
| XM_008763676.1 | XP_008761898.1 | 3 - 4 | 1239 | 54 | ||
| GenBank mRNA | EF187254.1 | - | 3 - 4 | 585 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car9
Genomic Map