Assay Details
| Gene Symbol: RGD1304595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106661.1 | NP_001100131.1 | 7 - 8 | 962 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1304595
Genomic Map