Assay Details
| Gene Symbol: Msrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031660.1 | NP_001026830.1 | 1 - 2 | 113 | 142 | |
| GenBank mRNA | BC098953.1 | - | 1 - 2 | 113 | 7374871 | 142 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC098953.1, NM_001031660.1 transcripts.
Set Membership: |
|

Msrb2
Genomic Map