Assay Details
| Gene Symbol: Msrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031660.1 | NP_001026830.1 | 3 - 4 | 282 | 64 | |
| GenBank mRNA | BC098953.1 | - | 3 - 4 | 282 | 7374871 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Msrb2
Genomic Map