Assay Details
| Gene Symbol: Nlrp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106220.1 | NP_001099690.1 | 2 - 3 | 1880 | 127 | |
| XM_008758899.1 | XP_008757121.1 | 3 - 4 | 1969 | 127 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nlrp4a
Genomic Map