Assay Details
| Gene Symbol: Tmprss11e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770102.1 | XP_008768324.1 | 5 - 6 | 434 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11e| Gene Symbol: Tmprss11e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770102.1 | XP_008768324.1 | 5 - 6 | 434 | 72 | |
Set Membership: |
|