Assay Details
| Gene Symbol: Stx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758601.1 | XP_008756823.1 | 1 - 2 | 119 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008758601.1 transcripts.
Set Membership: |
|

Stx11
Genomic Map