Assay Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005889.2 | NP_001005889.2 | 12 - 13 | 1798 | 62 | |
| GenBank mRNA | AY428868.1 | - | 11 - 12 | 1623 | 62 | |
| BC087147.1 | - | 12 - 13 | 1798 | 7130553 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P090479 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P385117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72522 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090479 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P385117 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090479 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P385117 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P385117 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P090479 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P146200 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P385117 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88174 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15010 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47452 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10909971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909973 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10909970 | - | - |
| Rat Genome 230 2.0 Array | 1396207_at | - | - |
| Rat Genome 230 2.0 Array | 1383825_at | - | - |
| Rat Genome 230 2.0 Array | 1396207_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379641_at | - | - |
| Rat Genome 230 2.0 Array | 1375542_at | - | - |
| Rat Genome 230 2.0 Array | 1378287_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379641_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383825_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375542_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378287_at | - | - |
| Rat Genome 230 2.0 Array | 1388619_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388619_at | - | - |
| Affymetrix HT Rat Focus Array | 1396207_at | - | - |
| Affymetrix HT Rat Focus Array | 1383825_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378287_at | - | - |
| Affymetrix HT Rat Focus Array | 1396207_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378287_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383825_at | - | - |
| Affymetrix HT Rat Focus Array | 1379641_at | - | - |
| Affymetrix HT Rat Focus Array | 1388619_at | - | - |
| Affymetrix HT Rat Focus Array | 1388619_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375542_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379641_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375542_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800004647.rn.1 | TC0800000699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800004648.rn.1 | TC0800000699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800004649.rn.1 | TC0800000699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800004646.rn.1 | TC0800000699.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388619_PM_at | - | - |
| Rat Expression Set 230 | 1388619_at | - | - |
| Rat Expression Set 230 | 1375542_PM_at | - | - |
| Rat Expression Set 230 | 1375542_at | - | - |
| Rat Expression Set 230 | 1396207_at | - | - |
| Rat Expression Set 230 | 1383825_at | - | - |
| Rat Expression Set 230 | 1396207_PM_at | - | - |
| Rat Expression Set 230 | 1379641_at | - | - |
| Rat Expression Set 230 | 1378287_PM_at | - | - |
| Rat Expression Set 230 | 1379641_PM_at | - | - |
| Rat Expression Set 230 | 1383825_PM_at | - | - |
| Rat Expression Set 230 | 1378287_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909970 | - | - |
| Rat Genome U34 Set | rc_AA944157_at | - | - |
| Rat Genome U34 Set | rc_AI230947_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22070300 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363289 | - | - |
| RatRef-12 Expression BeadChip | ILMN_55683 | - | - |

Rdx
Genomic Map