Assay Details
Gene Symbol: LOC102547257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242860.2 | XP_006242922.1 | 1 - 2 | 2376 | 80 |
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053519.1 | NP_445971.1 | 6361 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to Golgi
protein targeting
protein targeting to lysosome
post-Golgi vesicle-mediated transport
cell proliferation
regulation of smooth muscle cell migration
cell migration
negative regulation of protein binding
negative regulation of protein oligomerization
negative regulation of MAP kinase activity
protein retention in Golgi apparatus
positive regulation of protein catabolic process
negative regulation of neurogenesis
protein maturation
positive regulation of protein exit from endoplasmic reticulum
negative regulation of neuron death
negative regulation of beta-amyloid formation
positive regulation of choline O-acetyltransferase activity
negative regulation of tau-protein kinase activity
positive regulation of ER to Golgi vesicle-mediated transport
positive regulation of early endosome to recycling endosome transport
negative regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
positive regulation of protein localization to early endosome
negative regulation of neurofibrillary tangle assembly
positive regulation of endocytic recycling
beta-amyloid binding
low-density lipoprotein particle binding
ADP-ribosylation factor binding
protein targeting to Golgi
protein targeting
protein targeting to lysosome
post-Golgi vesicle-mediated transport
cell proliferation
regulation of smooth muscle cell migration
cell migration
negative regulation of protein binding
negative regulation of protein oligomerization
negative regulation of MAP kinase activity
protein retention in Golgi apparatus
positive regulation of protein catabolic process
negative regulation of neurogenesis
protein maturation
positive regulation of protein exit from endoplasmic reticulum
negative regulation of neuron death
negative regulation of beta-amyloid formation
positive regulation of choline O-acetyltransferase activity
negative regulation of tau-protein kinase activity
positive regulation of ER to Golgi vesicle-mediated transport
positive regulation of early endosome to recycling endosome transport
negative regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
positive regulation of protein localization to early endosome
negative regulation of neurofibrillary tangle assembly
positive regulation of endocytic recycling
beta-amyloid binding
low-density lipoprotein particle binding
ADP-ribosylation factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P552341 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P986792 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P986792 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P552341 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P552341 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P986792 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P986792 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P552341 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P250291 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P986792 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P552341 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10916529 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916520 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916523 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916528 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916522 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916525 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916524 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916521 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10916526 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10916493 | - | - |
Rat Genome 230 2.0 Array | 1377457_a_at | - | - |
Rat Genome 230 2.0 Array | 1393626_at | - | - |
Rat Genome 230 2.0 Array | 1394786_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394786_at | - | - |
Rat Genome 230 2.0 Array | 1386268_at | - | - |
Rat Genome 230 2.0 Array | 1377457_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1386269_PM_x_at | - | - |
Rat Genome 230 2.0 Array | 1390710_x_at | - | - |
Rat Genome 230 2.0 Array | 1390710_PM_x_at | - | - |
Rat Genome 230 2.0 Array | 1377458_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386269_x_at | - | - |
Rat Genome 230 2.0 Array | 1386268_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393626_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377458_at | - | - |
Rat Genome 230 2.0 Array | 1393933_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393933_at | - | - |
Affymetrix HT Rat Focus Array | 1390710_x_at | - | - |
Affymetrix HT Rat Focus Array | 1393933_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377457_a_at | - | - |
Affymetrix HT Rat Focus Array | 1386269_PM_x_at | - | - |
Affymetrix HT Rat Focus Array | 1390710_PM_x_at | - | - |
Affymetrix HT Rat Focus Array | 1386269_x_at | - | - |
Affymetrix HT Rat Focus Array | 1377458_at | - | - |
Affymetrix HT Rat Focus Array | 1393933_at | - | - |
Affymetrix HT Rat Focus Array | 1386268_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386268_at | - | - |
Affymetrix HT Rat Focus Array | 1377457_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1377458_PM_at | - | - |
Rat Expression Set 230 | 1390710_x_at | - | - |
Rat Expression Set 230 | 1386268_at | - | - |
Rat Expression Set 230 | 1377457_PM_a_at | - | - |
Rat Expression Set 230 | 1393933_at | - | - |
Rat Expression Set 230 | 1390710_PM_x_at | - | - |
Rat Expression Set 230 | 1386269_PM_x_at | - | - |
Rat Expression Set 230 | 1386268_PM_at | - | - |
Rat Expression Set 230 | 1377458_at | - | - |
Rat Expression Set 230 | 1377458_PM_at | - | - |
Rat Expression Set 230 | 1386269_x_at | - | - |
Rat Expression Set 230 | 1377457_a_at | - | - |
Rat Expression Set 230 | 1393933_PM_at | - | - |
Rat Expression Set 230 | 1393626_PM_at | - | - |
Rat Expression Set 230 | 1394786_at | - | - |
Rat Expression Set 230 | 1394786_PM_at | - | - |
Rat Expression Set 230 | 1393626_at | - | - |
Rat Genome U34 Set | rc_AA850618_at | - | - |
Rat Genome U34 Set | rc_AI177589_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22137784 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365362 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363716 | - | - |
RatRef-12 Expression BeadChip | ILMN_58491 | - | - |
