Assay Details
| Gene Symbol: Ostm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029925.1 | NP_001025096.1 | 444 | 87 | ||
| GenBank mRNA | BC097978.1 | - | 444 | 7442765 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ostm1
Genomic Map