Assay Details
| Gene Symbol: Clic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009651.1 | NP_001009651.1 | 3 - 4 | 504 | 129 | |
| GenBank mRNA | BC088182.1 | - | 3 - 4 | 504 | 7365286 | 129 |
| FQ227497.1 | - | 3 - 4 | 456 | 129 | ||
| FQ231721.1 | - | 3 - 4 | 456 | 129 | ||
| FQ234033.1 | - | 3 - 4 | 502 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clic2
Genomic Map