Assay Details
| Gene Symbol: LOC103692307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763581.1 | XP_008761803.1 | 2 | 617 | 68 | |
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170327.1 | NP_001163798.1 | 8 - 9 | 7003 | 68 | |
| XM_006248064.2 | XP_006248126.1 | 8 - 9 | 7021 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103692307
Genomic Map