Assay Details
| Gene Symbol: Stt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134749.1 | NP_001128221.1 | 15 - 16 | 1875 | 65 | |
| XM_006242808.1 | XP_006242870.1 | 15 - 16 | 1920 | 65 | ||
| GenBank mRNA | BC168223.1 | - | 15 - 16 | 1846 | 7324965 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Stt3a
Genomic Map