Assay Details
| Gene Symbol: Ccdc134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024355.1 | NP_001019526.1 | 3 - 4 | 515 | 101 | |
| XM_006242113.2 | XP_006242175.1 | 4 - 5 | 428 | 101 | ||
| GenBank mRNA | BC088206.1 | - | 3 - 4 | 515 | 7381354 | 101 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc134
Genomic Map