Assay Details
| Gene Symbol: Zfp706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126087.1 | NP_001119559.1 | 1 - 2 | 180 | 111 | |
| GenBank mRNA | BC083795.1 | - | 1 - 2 | 180 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC083795.1, NM_001126087.1 transcripts.
Set Membership: |
|

Zfp706
Genomic Map