Assay Details
| Gene Symbol: Kcns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023966.1 | NP_076456.1 | 1 - 2 | 168 | 107 | |
| GenBank mRNA | AJ296090.1 | - | 1 - 2 | 168 | 107 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ296090.1,NM_023966.1 transcripts.
Set Membership: |
|

Kcns2
Genomic Map