Assay Details
| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130582.2 | NP_001124054.2 | 3 - 4 | 643 | 62 | |
| XM_006242121.2 | XP_006242183.1 | 2 - 3 | 460 | 62 | ||
| GenBank mRNA | BC168759.1 | - | 3 - 4 | 643 | 6888763 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Xpnpep3
Genomic Map