Assay Details
| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130495.1 | NP_001123967.1 | 1 - 2 | 337 | 55 | |
| XM_006241931.2 | XP_006241993.1 | 1 - 2 | 349 | 55 | ||
| GenBank mRNA | BC168150.1 | - | 347 | 7457249 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift27
Genomic Map