Assay Details
| Gene Symbol: Zfp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142757.1 | NP_001136229.1 | 2 - 3 | 160 | 114 | |
| XM_006241927.2 | XP_006241989.1 | 2 - 3 | 314 | 114 | ||
| XM_006241928.2 | XP_006241990.1 | 2 - 3 | 314 | 114 | ||
| Gene Symbol: Znf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750369.3 | XP_003750417.2 | 2 - 3 | 314 | 114 | |
| XM_006241919.2 | XP_006241981.1 | 2 - 3 | 314 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142757.1,XM_003750369.3,XM_006241919.2,XM_006241927.2,XM_006241928.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp7
Genomic Map