Assay Details
| Gene Symbol: Znf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750369.3 | XP_003750417.2 | 4 - 5 | 554 | 61 | |
| XM_006241919.2 | XP_006241981.1 | 4 - 5 | 551 | 61 | ||
| XM_008765633.1 | XP_008763855.1 | 4 - 5 | 551 | 61 | ||
| Gene Symbol: Zfp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241927.2 | XP_006241989.1 | 4 - 5 | 554 | 61 | |
| XM_006241928.2 | XP_006241990.1 | 4 - 5 | 551 | 61 | ||
| XM_008765571.1 | XP_008763793.1 | 4 - 5 | 551 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Znf7
Genomic Map