Assay Details
| Gene Symbol: Fam135b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241723.2 | XP_006241785.1 | 4 - 5 | 754 | 61 | |
| XM_008765601.1 | XP_008763823.1 | 6 - 7 | 1143 | 61 | ||
More Information
Set Membership: |
|

Fam135b
Genomic Map