Assay Details
| Gene Symbol: Slc22a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013947.1 | NP_001013969.1 | 3 - 4 | 552 | 90 | |
| XM_008765502.1 | XP_008763724.1 | 3 - 4 | 520 | 90 | ||
| GenBank mRNA | AB559554.1 | - | 3 - 4 | 549 | 90 | |
| BC081982.1 | - | 3 - 4 | 552 | 7128584 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a22
Genomic Map