Assay Details
| Gene Symbol: Slc22a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013947.1 | NP_001013969.1 | 5 - 6 | 886 | 62 | |
| XM_008765502.1 | XP_008763724.1 | 5 - 6 | 854 | 62 | ||
| GenBank mRNA | AB559554.1 | - | 5 - 6 | 883 | 62 | |
| BC081982.1 | - | 5 - 6 | 886 | 7128584 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a22
Genomic Map