Assay Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024908.1 | NP_001020079.1 | 4 - 5 | 1200 | 79 | |
| XM_006241859.2 | XP_006241921.1 | 4 - 5 | 1689 | 79 | ||
| GenBank mRNA | BC097424.1 | - | 4 - 5 | 1200 | 7455079 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd3
Genomic Map