Assay Details
| Gene Symbol: Ybey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108529.1 | NP_001101999.1 | 3 - 4 | 448 | 75 | |
| XM_006256301.2 | XP_006256363.1 | 3 - 4 | 459 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ybey
Genomic Map