Assay Details
| Gene Symbol: RT1-DOb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008846.1 | NP_001008846.1 | 3 - 4 | 645 | 71 | |
| XM_006255970.2 | XP_006256032.1 | 3 - 4 | 769 | 71 | ||
| GenBank mRNA | KC222915.1 | - | 3 - 4 | 645 | 71 | |
| KC222930.1 | - | 3 - 4 | 567 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-DOb
Genomic Map