Assay Details
| Gene Symbol: RT1-DOb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008846.1 | NP_001008846.1 | 4 - 5 | 755 | 71 | |
| XM_006255970.2 | XP_006256032.1 | 4 - 5 | 879 | 71 | ||
| GenBank mRNA | KC222915.1 | - | 4 - 5 | 755 | 71 | |
| KC222916.1 | - | 4 - 5 | 755 | 71 | ||
| KC222930.1 | - | 4 - 5 | 677 | 71 | ||
| KC222931.1 | - | 4 - 5 | 677 | 71 | ||
More Information
Set Membership: |
|

RT1-DOb
Genomic Map