Assay Details
| Gene Symbol: Pole2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169108.1 | NP_001162579.1 | 6 - 7 | 513 | 65 | |
| XM_006240154.2 | XP_006240216.1 | 6 - 7 | 528 | 65 | ||
| XM_008764696.1 | XP_008762918.1 | 9 - 10 | 1312 | 65 | ||
| XM_008764697.1 | XP_008762919.1 | 8 - 9 | 1122 | 65 | ||
| XM_008764698.1 | XP_008762920.1 | 9 - 10 | 1251 | 65 | ||
| XM_008764699.1 | XP_008762921.1 | 9 - 10 | 1335 | 65 | ||
| XM_008764700.1 | XP_008762922.1 | 9 - 10 | 1221 | 65 | ||
| XM_008764701.1 | XP_008762923.1 | 9 - 10 | 1115 | 65 | ||
| XM_008764702.1 | XP_008762924.1 | 9 - 10 | 1211 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pole2
Genomic Map