Assay Details
| Gene Symbol: Sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271222.1 | NP_001258151.1 | 2 - 3 | 403 | 107 | |
| GenBank mRNA | FM089942.1 | - | 2 - 3 | 368 | 107 | |
| FM108041.1 | - | 2 - 3 | 403 | 107 | ||
| FM109760.1 | - | 2 - 3 | 355 | 107 | ||
| FQ221139.1 | - | 2 - 3 | 264 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sf3b6
Genomic Map