Assay Details
| Gene Symbol: Osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239886.2 | XP_006239948.1 | 1 - 2 | 300 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239886.2 transcripts.
Set Membership: |
|

Osr1
Genomic Map