Assay Details
| Gene Symbol: Slc4a1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106709.2 | NP_001100179.2 | 5 - 6 | 1183 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc4a1ap| Gene Symbol: Slc4a1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106709.2 | NP_001100179.2 | 5 - 6 | 1183 | 67 | |
Set Membership: |
|