Assay Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750123.2 | XP_003750171.1 | 3 - 4 | 368 | 67 | |
| XM_006239855.1 | XP_006239917.1 | 3 - 4 | 368 | 67 | ||
| XM_008764544.1 | XP_008762766.1 | 3 - 4 | 434 | 67 | ||
| XM_008764545.1 | XP_008762767.1 | 3 - 4 | 434 | 67 | ||
| XM_008764546.1 | XP_008762768.1 | 3 - 4 | 434 | 67 | ||
| GenBank mRNA | FQ232123.1 | - | 1 - 2 | 331 | 67 | |
| FQ233391.1 | - | 1 - 2 | 331 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cenpo
Genomic Map