Assay Details
| Gene Symbol: Fam228a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014074.1 | NP_001014096.1 | 3 - 4 | 426 | 100 | |
| XM_006239858.2 | XP_006239920.1 | 3 - 4 | 718 | 100 | ||
| GenBank mRNA | BC083643.1 | - | 3 - 4 | 426 | 7134934 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam228a
Genomic Map