Assay Details
| Gene Symbol: Sypl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014263.1 | NP_001014285.1 | 2 - 3 | 186 | 61 | |
| XM_006240001.2 | XP_006240063.1 | 2 - 3 | 199 | 61 | ||
| XM_006240002.2 | XP_006240064.1 | 2 - 3 | 199 | 61 | ||
| GenBank mRNA | BC082078.1 | - | 2 - 3 | 186 | 7133263 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Sypl1
Genomic Map