Assay Details
| Gene Symbol: Ppm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107681.2 | NP_001101151.1 | 3 - 4 | 785 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P489817 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P170939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71485 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P489817 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P170939 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P489817 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P170939 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P489817 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P170939 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P288925 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P170939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7497 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100216 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99862 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44793 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16986 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95732 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21203 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46756 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10815922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815921 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10815919 | - | - |
| Rat Genome 230 2.0 Array | 1376771_at | - | - |
| Rat Genome 230 2.0 Array | 1375606_at | - | - |
| Rat Genome 230 2.0 Array | 1376584_at | - | - |
| Rat Genome 230 2.0 Array | 1376584_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376771_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375606_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200008527.rn.1 | TC0200001366.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200008528.rn.1 | TC0200001366.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200008529.rn.1 | TC0200001366.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376771_PM_at | - | - |
| Rat Expression Set 230 | 1376584_PM_at | - | - |
| Rat Expression Set 230 | 1376771_at | - | - |
| Rat Expression Set 230 | 1376584_at | - | - |
| Rat Expression Set 230 | 1375606_at | - | - |
| Rat Expression Set 230 | 1375606_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815919 | - | - |
| Rat Genome U34 Set | rc_AI235906_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22324316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366202 | - | - |

Ppm1l
Genomic Map