Assay Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203340.1 | NP_976085.1 | 5 - 6 | 498 | 96 | |
| XM_006252261.2 | XP_006252323.1 | 6 - 7 | 1308 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npm2
Genomic Map