Assay Details
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107943.1 | NP_001101413.1 | 5 - 6 | 729 | 59 | |
| XM_006238363.2 | XP_006238425.1 | 5 - 6 | 771 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cell-matrix adhesion
ceramide metabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
biological_process
positive regulation of cell proliferation
positive regulation of cell death
response to retinoic acid
negative regulation of cell adhesion mediated by integrin
cellular response to drug
sphingosine biosynthetic process
negative regulation of protein glycosylation in Golgi
molecular_function
ceramidase activity
dihydroceramidase activity
negative regulation of cell-matrix adhesion
ceramide metabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
biological_process
positive regulation of cell proliferation
positive regulation of cell death
response to retinoic acid
negative regulation of cell adhesion mediated by integrin
cellular response to drug
sphingosine biosynthetic process
negative regulation of protein glycosylation in Golgi
molecular_function
ceramidase activity
dihydroceramidase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P674018 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P115566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54164 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P115566 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P674018 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P674018 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P115566 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P115566 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P674018 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P269925 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100524 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10869813 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869812 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869809 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869810 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869811 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10869808 | - | - |
| Rat Genome 230 2.0 Array | 1379846_at | - | - |
| Rat Genome 230 2.0 Array | 1391791_at | - | - |
| Rat Genome 230 2.0 Array | 1373302_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391791_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379846_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373302_at | - | - |
| Affymetrix HT Rat Focus Array | 1373302_at | - | - |
| Affymetrix HT Rat Focus Array | 1391791_at | - | - |
| Affymetrix HT Rat Focus Array | 1391791_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373302_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500005960.rn.1 | TC0500000919.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500005961.rn.1 | TC0500000919.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500005962.rn.1 | TC0500000919.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373302_PM_at | - | - |
| Rat Expression Set 230 | 1373302_at | - | - |
| Rat Expression Set 230 | 1379846_PM_at | - | - |
| Rat Expression Set 230 | 1391791_at | - | - |
| Rat Expression Set 230 | 1379846_at | - | - |
| Rat Expression Set 230 | 1391791_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869808 | - | - |
| Rat Genome U34 Set | rc_AA848826_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22117728 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364753 | - | - |

Acer2
Genomic Map