Assay Details
Gene Symbol: LOC501038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047965.1 | NP_001041430.1 | 2 - 3 | 163 | 107 | |
XM_006243552.2 | XP_006243614.1 | 2 - 3 | 1690 | 107 | ||
GenBank mRNA | AY325184.1 | - | 2 - 3 | 163 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
