Assay Details
| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168641.1 | NP_001162112.1 | 1 - 2 | 102 | 74 | |
| XM_006233028.2 | XP_006233090.1 | 1 - 2 | 2046 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wars2
Genomic Map