Assay Details
| Gene Symbol: Edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191899.1 | NP_001178828.1 | 5 - 6 | 523 | 64 | |
| XM_006256381.2 | XP_006256443.1 | 6 - 7 | 816 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Edar
Genomic Map