Assay Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034946.1 | NP_001030118.1 | 3 - 4 | 63 | ||
| GenBank mRNA | BC099766.1 | - | 3 - 4 | 7931002 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itln1
Genomic Map