Assay Details
| Gene Symbol: Plscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012139.2 | NP_001012139.1 | 5 - 6 | 738 | 73 | |
| XM_008767838.1 | XP_008766060.1 | 4 - 5 | 1089 | 73 | ||
| XM_008767839.1 | XP_008766061.1 | 4 - 5 | 2073 | 73 | ||
| GenBank mRNA | AY548831.1 | - | 4 - 5 | 615 | 73 | |
| BC098055.1 | - | 5 - 6 | 738 | 7457695 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plscr3
Genomic Map