Assay Details
| Gene Symbol: Gpr161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751260.3 | XP_003751308.1 | 2 - 3 | 435 | 85 | |
| XM_006250191.2 | XP_006250253.1 | 2 - 3 | 505 | 85 | ||
| XM_008769710.1 | XP_008767932.1 | 3 - 4 | 516 | 85 | ||
| XM_008769711.1 | XP_008767933.1 | 2 - 3 | 454 | 85 | ||
| XM_008769712.1 | XP_008767934.1 | 2 - 3 | 433 | 85 | ||
More Information
Set Membership: |
|

Gpr161
Genomic Map