Assay Details
| Gene Symbol: Ankrd66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244618.2 | XP_006244680.1 | 4 - 5 | 880 | 59 | |
| XM_008766912.1 | XP_008765134.1 | 4 - 5 | 877 | 59 | ||
| XM_576520.4 | XP_576520.3 | 3 - 4 | 1154 | 59 | ||
More Information
Set Membership: |
|

Ankrd66
Genomic Map