Assay Details
| Gene Symbol: Ap1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172150.1 | NP_001165621.1 | 2 - 3 | 303 | 121 | |
| XM_006245264.2 | XP_006245326.1 | 2 - 3 | 356 | 121 | ||
| XM_008767284.1 | XP_008765506.1 | 2 - 3 | 353 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap1s3
Genomic Map