Assay Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001071401.3 | XP_001071401.1 | 2 - 3 | 756 | 67 | |
| XM_006243870.2 | XP_006243932.1 | 2 - 3 | 1902 | 67 | ||
| XM_008766597.1 | XP_008764819.1 | 3 - 4 | 482 | 67 | ||
More Information
Set Membership: |
|

Lsmem2
Genomic Map