Assay Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001071401.3 | XP_001071401.1 | 3 - 4 | 943 | 60 | |
| XM_006243870.2 | XP_006243932.1 | 3 - 4 | 2089 | 60 | ||
| XM_008766597.1 | XP_008764819.1 | 4 - 5 | 669 | 60 | ||
More Information
Set Membership: |
|

Lsmem2
Genomic Map