Assay Details
| Gene Symbol: Gbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233425.2 | XP_006233487.1 | 3 - 4 | 475 | 60 | |
| XM_008761538.1 | XP_008759760.1 | 4 - 5 | 670 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gbp1
Genomic Map